Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLview™ Sterile Cell Culture Dishes With Glass Bottom

Free of detectable DNase, RNase, human DNA. Greiner Bio-One CELLview™ Cell Culture Dishes With Glass Bottom features high transparent achromatic borosilicate glass; hydrolytic class 1 (DIN-ISO 719).

Supplier:  Greiner Bio-One 627861

 View more versions of this product

Catalog No. 07-000-233


Add to Cart
This item is not returnable. View return policy

Description

Description

  • Non-pyrogenic
  • Non-cytotoxic
  • Free of detectable DNase, RNase, human DNA
  • High transparent achromatic borosilicate glass
  • Maximal spectral transmission, no autofluorescence, no depolarisation of light
  • Available

    Certifications

    ISO 8255-1:1986

    TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Polystyrene
Flat
Clear
Single-use
Round
Non-treated
5 mL
Glass bottom thickness: 175 μm +/- 15 μm
35 mm
Closure Included
One compartment
10 mL
8.7 sq. cm
Polystyrene
Sterile
Cell Culture Dish
Lid
10 mm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit