Learn More
Advantec MFS, Inc Cellulose Extraction Thimble Filters No 84, 25X60mm
Supplier: Advantec MFS, Inc N08425X29X60MM
Cellulose Extraction Thimble Filters made of high-purity alpha cotton are seamless for strength. Analyze fats, oils, grease, and pollutants. Soxlet extractions of organic components. Nominal retention 8um; OD 29mm. (Other sizes available)