Learn More
Cellpath CellSafe 5 Core Biopsy Cassettes with Five Wells
Reduces the pressure on laboratory staff by enabling them to perform five core biopsies in numbered wells sectioned together for quick, cost effective preparation. CellPath CellSafe Biopsy cassettes are used for breast and prostrate samples.
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-222-004
|
Cellpath
EBG020602A |
Blue | N/A | N/A | |||||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
22-222-003
|
Cellpath
EBG020102A |
White | Item Discontinued This product is either new or inactive, and cannot be ordered online.This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Features integral X-ray lucent fine mesh for efficient processing
- Simultaneous preparation of core biopsies
- Core biopsies for breast and prostate samples
- Each well is numbered for identification
- Available in two colors (white and blue)
- Blue cassette produced to aid contrast and visualization of white or light colored biopsies
Specifications
100/Pk. |