Learn More
Greiner Bio-One CELLCOAT™ 384-well, Collagen Type I-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Non-sterile; Lid:With Lid
Supplier:  Greiner Bio-One 781950
Catalog No. 07-000-898
Add to Cart
Description
Description
  • Promotes growth of endothelial cell, hepatocytes, muscle cells, pheochromocytoma cells (PC12) and other cell types
  • Cell cultivation in serum-free or serum-reduced medium
  • Rat tail collagen type I
  • Increase in isolation and cu
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
16 x 24
Clear
384
Non-sterile
131 μL
4800 x g (swinging bucket rotor)
5
With Lid
Not Autoclavable
Polystyrene
15 to 110 μL
Collagen Type I
Rounded Square
20/Cs.
Room Temperature
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
ShelfLife : 24 Months after month of production