Learn More
Andwin Scientific CELL SPREADER 500/PK INDIVWRAP
SDP
Supplier:  Andwin Scientific 47732760
Encompass_Preferred
The polystyrene spreaders feature a unique L-shaped design that permits smooth and uniform sample distribution and decreases the chance of the spreader digging into the agar. The disposable and nonbreakable spreaders are efficient alternative to bending glass rods or pipets. This product is sold in a pack of 500 individually wrapped spreaders.
Catalog No. NC0385299
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »