Learn More
Foxx Life Sciences Abdos Cell Spreader Triangle Shaped, Gamma Radiated Polypropylene (PP) Blue with a Length 140mm, Individually Wrapped, 400/CS
Supplier:  Foxx Life Sciences P10991
Encompass_Preferred
Abdos Cell Spreaders are made of high-quality Polypropylene (PP) and are designed to spread and disperse liquid onto the surface of an agar plate without gouging or cutting the medium and to allow for maximum recovery.Individually WrappedGamma RadiatedSterile Plates have been validated according to EN ISO 11137-2; 2012 for SAL 10-6In a highly anticipated collaboration, Foxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. Abdos follows intense quality control measures, audits, and are ISO 9001:2015 and ISO 13485:2016 certified company and CE marked. Manufactured on state-of-the-art machines with precise molds and virgin medical grade resin, these outstanding products are a valuable addition to any workspace. For downloadable certifications and Certificates of Analysis please visit: http://www.abdoslifesciences.com/Global/certificates.html.
Catalog No. NC1819205