Learn More
Sigma Aldrich Fine Chemicals Biosciences CryoStor cell cryopreservation media, 100 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences C2874100ML
CryoStor, a series of cell-specific, optimized preservation media, is uniquely formulated to address the molecularbiological aspects of cells during the cryopreservation process thereby directly reducing the level of Cryopreservation-Induced Delayed-Onset Cell Death and improving post-thaw cell viability and function.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.