Learn More
Celestron™ Skyportal WiFi Module
SCI_ED
For aligning and controlling telescope wirelessly using smartphone or tablet.
Supplier:  Celestron™ 93973
Catalog No. S13587
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
Description
Description
  • Plug into telescope’s Hand Control or AUX port and connect to wireless signal with device
  • Tap any celestial object that is seen in sky to identify it instantly
  • Tap again and telescope slews to that object, centering it perfectly in eyepiece
  • Explore 100,000 objects in SkyPortal’s database
  • Can observe, view astroimages and listen to audio descriptions for hundreds of most popular objects including planets, Orion Nebula, Hercules Open Star Cluster
  • Sky Tour feature generates list of all best stars, galaxies and nebulae to view based on your exact time and location
  • GPS coordinates not required
  • Unit does not require batteries or external power
  • Compatible with following Celestron mounts: Advanced VX; CG-5 (requires compatible cable); CGE Pro; CGEM; CGEM DX; CPC; LCM; NexStar Evolution; NexStar SLT; NexStar SE; SkyProdigy (no automatic alignment)
Warranty and Services
2 years
Specifications
Specifications
Skyportal WiFi Module
2 Years