Learn More
Celestron™ Motor Drive for the AstroMaster/PowerSeeker Telescopes
SCI_ED
For Celestron CG-2 & CG-3 equatorial mounts.
Supplier:  Celestron™ 93514
Catalog No. S98228K
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
  • Right Ascension (RA) tracking compensates for the Earth's rotation
  • Equipped with a North/South switch that can reverse the direction of the motor for Southern Hemisphere use
  • Minor adjustments in Declination keep celestial objects centered in the eyepiece for long periods of time
  • Adjustable motor speed
  • Runs on 9V battery
Warranty and Services
Two years
Specifications
Specifications
2 Years
Computerized Equatorial