Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ cDNA Synthesis & Go Kit
Provides a rapid and very sensitive method for first strand cDNA synthesis for use in real-time PCR studies
Supplier: MP Biomedicals™ 11EBI00005
Description
- cDNA Synthesis& Go Kit provides a rapid and very sensitive method for first strand cDNA synthesis for use in real-time PCR studies.
- The 5x specific Buffer provides highly optimized components for efficient reverse transcription, and includes a unique blend of anchored oligo dT and random hexamer primers to ensure unbiased 3' and 5' coverage for enhanced data accuracy.
- An extremely efficient reverse transcriptase delivers highly robust first strand synthesis and higher cDNA yields from a wide range of input RNA concentrations.
- cDNA Synthesis& Go Kit offers enhanced sensitivity, efficiency and reproducibility for exceptional performance in subsequent real-time PCR experiments.
Specifications
Store at -20°C | |
50 U | |
Gene Expression, Pathogen Detection, miRNA analysis |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?
Provide Content Correction