Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD43 Rat anti-Mouse, RB780, Clone: S7, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 755808
This item is not returnable.
View return policy
Description
CD43 Monoclonal antibody specifically detects CD43 in Mouse samples. It is validated for Flow CytometrySpecifications
CD43 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
S7 | |
RB780 | |
Spn; Sialophorin; Leukosialin; Ly-48; Ly48; Galgp; LEUK | |
Mouse Plasmacytoma MOPC-315 | |
RUO | |
20737 | |
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title