Learn More
CD41 Rat anti-Mouse, Oligo, Clone: MWReg30, BD™ AbSeq ™
Rat Monoclonal Antibody
Supplier:  BD Biosciences 940155
Catalog No. BDB940155
Add to Cart
This item is not returnable. View return policy
Description
Description
CD41 Monoclonal antibody specifically detects CD41 in Mouse samples. It is validated for Single Cell 3' Sequencing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD41
Monoclonal
Oligo
Itga2b; Integrin alpha 2b; Integrin alpha-IIb; Integrin α IIb; GpIIb
Mouse Platelets
RUO
Mouse
IgG1 κ
Single Cell 3' Sequencing
Clone MWReg30
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions.
Product Suggestions
Product Suggestions