Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Mouse anti-Rat, RB780, Clone: OX-34, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 755880
This item is not returnable.
View return policy
Description
CD2 Monoclonal antibody specifically detects CD2 in Rat samples. It is validated for Flow CytometrySpecifications
CD2 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Flow Cytometry | |
OX-34 | |
RB780 | |
LFA-2; OX34; Cd2 | |
Rat T blasts from mixed lymphocyte reactions | |
RUO | |
Rat | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title