Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Mouse anti-Human, Oligo, Clone: SJ25C1 (also known as SJ25-C1), BD™ AbSeq ™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 940004
This item is not returnable.
View return policy
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Single Cell 3' SequencingSpecifications
CD19 | |
Monoclonal | |
Oligo | |
B4; B-lymphocyte antigen CD19; Leu-12 | |
Human NALM1 + NALM16 cells | |
RUO | |
12483 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Single Cell 3' Sequencing | |
Clone SJ25C1 (also known as SJ25-C1) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction