Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD13 Mouse anti-Human, RB780, Clone: L138 (also known as Leu-M7), BD OptiBuild™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 755811

Catalog No. BDB755811


This item is not returnable. View return policy

Description

Description

CD13 Monoclonal antibody specifically detects CD13 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD13
Monoclonal
0.2 mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Human
IgG1 κ
Flow Cytometry
L138 (also known as Leu-M7)
RB780
ANPEP; APN; Aminopeptidase N; Alanyl aminopeptidase; LAP1; PEPN
KG-1a Cell Line
RUO
290
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit