Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11a Mouse anti-Human, RB780, Clone: G25.2 (also known as G-25.2), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 755973
This item is not returnable.
View return policy
Description
CD11a Monoclonal antibody specifically detects CD11a in Human samples. It is validated for Flow CytometrySpecifications
CD11a | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Flow Cytometry | |
G25.2 (also known as G-25.2) | |
RB780 | |
ITGAL; LFA-1 alpha; LFA-1A; LFA1A; integrin alpha L | |
Human Cytotoxic T Lymphocytes | |
RUO | |
Human | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title