Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCG 63802, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Reversible inhibitor of regulator of G-protein signaling (RGS) proteins. Selective amongst RGS proteins, with greatest potency at RGS4. Inhibits GTPase accelerating protein activity of RGS4 and blocks its interaction with Gαo. Retains activity under reducing conditions.
Specifications
Quantity | 50 mg |
Formula Weight | 450.51 |
Percent Purity | >99% |
Chemical Name or Material | CCG 63802 |
Safety and Handling
Recommended Storage : Store at +4°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title