Learn More
Edwards Fittings for BOC High Vacuum Pumps: Elbow Joint
For use with BOC high vacuum pumps
Supplier: Edwards C10514410
Description
- Made of aluminum
- Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
Specifications
BOC High Vacuum Pump | |
Aluminum | |
50 mm | |
NW25 Elbow joint |