Learn More
SKC Inc CASSETTE BLCK POLYP 25MM 50/PK
SDP
Supplier:  SKC Inc 225329
Encompass
SKC 3-section, 25 mm, black polypropylene filter cassette blank (Goblet style) with 1/2 in. spacer, conductive, (pkg/50).
Catalog No. NC0190857