Learn More
Carolina™ Sea Urchin Embryology Set
Students can watch the fascinating life cycle of sea urchins right in your classroom.
Supplier: Carolina Biological Supply 162500
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes:
- 14 fertile urchins (either Eucidaris tribuloides or Lytechinus variegatus)
Description
We guarantee at least 50% of the urchins to be ripe on arrival and recommend that they be used the day of arrival or immediately the next day to avoid reabsorption of gametes.
- Follow development from early cleavage through pluteus
- Not sexed, but the male-to-female ratio is typically about 1:1
Specifications
Students can watch the fascinating life cycle of sea urchins right in the classroom. | |
14/Set | |
14 fertile urchins (either Eucidaris tribuloides or Lytechinus variegatus), follow development from early cleavage through pluteus, not sexed, but the male-to-female ratio is typically about 1:1 | |
Sea Urchin Embryology Set |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.