Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Elodea Tips
Use these young growing tips to demonstrate streaming protoplasm as well as osmosis, plasmolysis, and other cell studies.
Supplier: Carolina Biological Supply 157340
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
- Excellent choice for studying plant cells, osmosis, and plasmolysis
- Great source for photosynthetic parenchyma cells
- Can be used to teach plant leaf morphology
- Not for aquarium use
- For a class of 15
Specifications
Used to demonstrate streaming protoplasm as well as osmosis, plasmolysis and other cell studies, for a class of 15, teach plant leaf morphology. | |
Contains 6 to 8 Elodea tips | |
Elodea Tip |
Ea. | |
Class of 15 students |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction