Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Earthworms
SDPSCI_ED

Students can find these tube-shaped, segmented animals living in soil and feeding on live and dead organic matter.

Specifications

For Use With (Application) Laboratory study and dissection
Storage Requirements 15°C
Size Large
Scientific Name Lumbricus terrestris
Includes Instructions
View More Specs

Alert:

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000


Products 3
Catalog Number Mfr. No. Product Type Quantity Size For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Size For Use With (Application) Price Quantity & Availability  
S07603ND
SDPSCI_ED
View Documents
Carolina Biological Supply
141620
Earthworm 12/Container Large Laboratory study and dissection N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
S07604ND
SDPSCI_ED
View Documents
Carolina Biological Supply
141624
Earthworm 100/Container Large Laboratory study and dissection N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
S07271ND
SDPSCI_ED
View Documents
Carolina Biological Supply
141622
Earthworm 25/Container Large Laboratory study and dissection N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent.
Description

Description

  • Lumbricus terrestris
  • Large, mature specimens are suitable for laboratory study and dissection
  • Medium-sized specimens are not suitable for dissection, but are good for making slides and for use as food for snakes and large salamanders
  • With instructions

Specifications

Specifications

Laboratory study and dissection
Large
Instructions
15°C
Lumbricus terrestris
Earthworm
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit