Learn More
Thermo Scientific CARBOPAC PA100 HPLC 4X250
Supplier: Thermo Scientific 043055
CarboPac PA100 Analytical Column (4 x 250 mm). 100% compatible with common organic solvents; ionic form eluents: sodium acetate and sodium hydroxide only.