Learn More
Thermo Scientific CARBOPAC MA1 250X4MM COLUMN
Supplier: Thermo Scientific 044066
For sensitive, rugged, and reliable separations of reduced sugars without the need for derivatization, use Thermo Scientific(TM) Dionex(TM) CarboPac(TM) MA1 Analytical & Guard Columns. These high-capacity, strong anion-exchange columns efficiently separate reduced monosaccharides and disaccharides commonly found in food products and physiological samples. Achieve baseline resolution of fucose, N-acetyl-(D)-glucosamine, N-acetyl-galactosamine, mannose, glucose, galactose, and neutral oligosaccharides in the same separation. Diameter (Metric):4mm; Flow Rate: 0.2 to 0.5mL/min;Max. Pressure:2000psi;Mobile Phase Compatibility:Incompatible with solvents and anionic detergent; Hydroxide eluents only; Particle Size:7.5um;pH: 0-14