Learn More
Parker Carbon (Odor) Filter
The addition of an odor removing carbon filter allows the CRYSTAL-AIRE™uv to retain odors, and to absorb gaseous materials by trapping them within the tiny pores of the activated carbon media.
Supplier: Parker 3310163AGS
Description
- Absorbs gaseous materials by trapping them within the tiny pores of the activated carbon media
- Filter should be replaced on a monthly basis
- Replacements are available in a pack of 12
Specifications
Activated Carbon | |
CRYSTAL-AIRE™uv System | |
1 Month |
Absorbing odors | |
Filter |