Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Disulfide (Laboratory), Fisher Chemical

Catalog Number C183212
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
75-15-0
CS2
76.131
MFCD00011321
QGJOPFRUJISHPQ-UHFFFAOYSA-N
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
6348
CHEBI:23012
methanedithione
C(=S)=S
75-15-0
CS2
76.131
MFCD00011321
QGJOPFRUJISHPQ-UHFFFAOYSA-N
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
6348
CHEBI:23012
methanedithione
C(=S)=S
Melting Point -111°C
Color Yellow
Boiling Point 46°C
Quantity 2.5 L
Identification Pass Test
Packaging Glass
Physical Form Liquid
Vapor Pressure 297.5mmHg at 20°C
Viscosity 0.363 cps at 20 deg C
Formula Weight 76.14
Grade Laboratory
Chemical Name or Material Carbon Disulfide
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit