Learn More
Cole-Parmer Glass Capillary Tubes, Closed Both Ends, 2.0 mm OD; 1000/PK
Supplier: Cole-Parmer 0301068
These capillary tubes are compatible with Electrothermal's digital melting point meters.