Learn More
Cadence Science™ Cancer Implant Needle
Needle provides simple procedure to obtain and implant tumorous tissue cells beneath skin of lab animals for observation while maintaining a sterile field
Supplier: Cadence Science™ 7928
Description
- Two piece needle set
- Consists of sharp pointed stainless-steel outer cannula and stainless steel blunt obturator which protrudes 3.2mm beyond the needle point
Specifications
Obtain and implant tumorous tissue cells beneath the skin of lab animals | |
Stainless Steel | |
11 ga | |
8.25 cm |
Cancer Implant Needle | |
Standard | |
3.25 in. |