Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GVS™ Calyx™ Capsule Filters
GVS™ Calyx™ Capsule Filters are all purpose polypropylene filters, designed for the removal of particles or bacteria from aqueous or solvent solutions and gas streams.
Supplier: GVS™ 1213596
Description
- Disposable filtration units containing membrane media, preflushed with purified water to reduce extractables
- Pass class VI toxicology testing and are integrity tested prior to shipment
- Ready to use, eliminating the need to disassemble, clean and reassemble filter housings
- Contain no glue or surfactants and feature serial layer filter design for Increased throughput and extended life
- Two upstream vents are included to facilitate venting in any position
- All purpose, designed for the removal of particles or bacteria from aqueous or solvent solutions and gas streams
- Food compliant (FDA/EU)
Specifications
Autoclavable | |
Polypropylene | |
Non-sterile | |
Polypropylene | |
80 psi @ 70°F (liquid); 50psi @ 70°F (Gas) | |
1 | |
43°C | |
9 cm |
Capsule Filter | |
0.45 μm | |
748 cm2 | |
1/4 in NPT male | |
1/4 in NPTM | |
110°F | |
3.5 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction