Learn More
Eisco™ Calcite Mineral Specimen
SCI_ED
Calcite Mineral Specimen is used in classroom identification labs, exercises, and lessons on chemical reactions.
Supplier:  Eisco™ ESNG0013
 View more versions of this product
Catalog No. S24683
Add to Cart
Description
Description
  • Standard in Moh's scale of hardness (3)
  • Example of three-plane rhombic cleavage
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Specifications
Mineral, Calcite
Clear labeled and barcoded bag
2 to 3 cm
Study on mineral hardness, cleavage, light refraction, and effervescence
1 Ea.
Product Suggestions
Product Suggestions