Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Protein G Plus/Protein A Agarose Suspension
Used in immunoprecipitation applications
Supplier: MilliporeSigma™ IP051.5ML
Description
- Blocked with BSA to reduce non-specific binding
- Best for bovine, goat, human, rat IgG for mouse IgG1
- Form: Liquid slurry
- Formulation: In PBS
- Preservative:<0.1% sodium azide
Specifications
2°C to 8°C | |
1.5 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction