Learn More
Cadence Science™ Micro-Emulsifying Needles
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
Supplier: Cadence Science™ 7978
Description
Double-hub needles are designed for use in mixing two liquid media differing in viscosity—especially for adjuvants and antigens, oil-based liquids, allergens
- Available with or without a reinforcing bar
- Overall length: 2 7/8 in. (7.3cm), except for 14-825-17K and -17L needles which have a length of 1 7/8 in. (4.8cm)
Specifications
Glass | |
For allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity, with reinforcing bar | |
1.87 in. | |
47.5 cm |
20 G | |
Without reinforcing bar | |
Micro Emulsifying Needle w/o Reinforcing Bar | |
Non-sterile |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.