Learn More
iQ Biosciences C57BL/6 Mouse Bone Marrow Cells, 20M cells/vial
Supplier: iQ Biosciences IQBMBM202
High-quality, high-viability (typically > 70%) bone marrow cells from the C57BL/6 mouse, the most commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including cell population characterization and differentiation into specific cell types. All orders come with an iQ Certificate of Analysis.