Learn More
Fine Science Tools Burrs for Micro Drill, Carbon Steel, 2.9 mm Tip Dia, 44 mm, 10/pk
Supplier: Fine Science Tools 1900729
Tip Diameter: 2.9mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Carbon Steel/ Length: 44mm/ Autoclave Safe: No