Learn More
Cole-Parmer Burkle 9695-1010 Titrating burette automatic, borosilicate glass; 10 ml
Supplier:  Cole-Parmer 0604446
Encompass_Preferred
These automic titrating burettes (burets) feature graduations according to Category B (tolerances according to DIN EN ISO 385). The included bottle, tube, and stopcock are made of polyethylene, polypropylene, natural rubber, and borosilicate glass wetted parts.
Catalog No. NC1633535