Learn More
SW Safety Bullseye Hi-Vis Cut-Resistant Level A5 TPR Gloves with AxiFybr
SDP
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Bullseye Hi-Vis Gloves with AxiFybr are equipped with a Velcro™ wrist closure, proprietary high flexibility TPR, padded palm, extended thumb crotch patch, and a distinct feature that allows for Stop & Go signaling at a distance. This tough glove holds up well against in high-risk environments.
  • 13-gauge AxiFybr engineered yarn liner delivers comfort at high levels of cut resistance.
  • Sandy microporous nitrile coating provides superior grip in oily and wet conditions.
  • Advanced flexible TPR for ANSI level 2 impact resistance that does not sacrifice mobility or comfort.
  • Palm padding to protect hands from repetitive impacts and vibrating tools.
Construction Knit
Coating Material Nitrile
Color Red, Yellow
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material AxiFybr
Size 3X-Large
Gender Unisex
Glove Cuff Style Knit
Liner Material AxiFybr
Model C562317
Series C56231
Yarn Gauge 13 G
Length (English) 10.2 in.