Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Replacement Filters for 50 CFM Clean Air Box Series Breathing Air Filtration Systems
SDP

Filter element replacement parts for use with 50 CFM clean air boxes.

$44.10 - $311.50

Specifications

For Use With (Equipment) CAB 50 cfm breathing air filtration system
Product Type Replacement Filter
View More Specs

Products 4
Catalog Number Mfr. No. Filter Efficiency Includes Price Quantity & Availability  
Catalog Number Mfr. No. Filter Efficiency Includes Price Quantity & Availability  
19-100-684
SDP
View Documents
Bullard™
CAB50FEA
Removes 95% of solid contaminants and bulk liquids, down to 5μ from the breathing air system Element A-Particulate
Each for $44.10
Add to Cart
 
19-100-685
SDP
View Documents
Bullard™
CAB50FEB
Removes particulates and extremely fine oil mists down to 0.01μ with 99.9% filter efficiency from the breathing air system Element B-Coalescing
Each for $140.70
Add to Cart
 
19-100-686
SDP
View Documents
Bullard™
CAB50FEC
Removes unpleasant oil and hydrocarbon vapors and odors from the breathing air system
Each for $160.80
Add to Cart
 
19-100-687
SDP
View Documents
Bullard™
CAB50FK
Removes 95% of solid contaminants, bulk liquids down to 5μ, particulates, extremely fine oil mists down to 0.01μ with 99.9% filter efficiency, unpleasant oil and hydrocarbon vapors and odors from the breathing air system Element A-Particulate, Element B-Coalescing, Element C-Activated Charcoal
Each for $311.50
Add to Cart
 
Description

Description

  • Replacement part for Bullard Clean Air Boxes (CAB) provide 3-stage filtration
  • For use with 50 CFM air flow CAB
  • Three stages of filtration remove oil, water, particulate, oil vapor and odors
  • CAB50FEA Stage 1 Element
  • CAB50FEB Stage 2 Element
  • CAB50FEC Stage 3 Element
  • CAB50FK Element kit, includes Stages 1-3
Specifications

Specifications

CAB 50 cfm breathing air filtration system
Replacement Filter
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit