Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GVS™ Buffer Swabs 4mL
Buffer swabs are used for collection of surface contamination from flat or convoluted surfaces prior to transport to a laboratory for culture and enumeration. Buffer swabs contain no bacteriostatic or bactericidal compounds and cannot suppress the action of detergents.
Supplier: GVS™ 10498305
Description
Description
- Used for the collection of surface contamination from flat or convoluted surfaces prior to transport to a laboratory for culture and enumeration. Buffer swabs contain no bacteriostatic or bactericidal compounds and cannot suppress the action of detergents.
Features and Benefits:
- Individually wrapped
- Qualitative and semi-quantitative hygiene control
- Sterile packed and ready-for-use
- Easy to handle
- Rapid results
- Long shelf-life
Applications:
- Used for the collection of surface contamination from flat or convoluted surfaces prior to transport to a laboratory for culture and enumeration.
The SwabCheck principle
- The surface is wiped with a cellulose swab and any bacteria collected are transferred via the swab into a tube containing a special medium with an indicator dye, which is then incubated.
- A single bacterium is sufficient to cause a color change.
- The SwabCheck is about 1000 times more sensitive than the conventional ATP method. This accuracy is particularly important in the food industry. With this simple method, it is possible to identify microorganisms such as Listeria monocytogenes, which must not be present in any concentration in food and beverages.
Specifications
Cellulose | |
Cellulose | |
125 |
Sterile | |
Buffer swabs used for collection of surface contamination |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Buffer swabs contain no bacteriostatic or bactericidal compounds and cannot suppress the action of detergents.
Spot an opportunity for improvement?
Provide Content Correction