Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pH Buffer Salt, pH 7.41, in ready to mix packets, Certified, Fisher Chemical

Each packet (envelope) contains sufficient material to prepare 1 liter of solution with a pH of 7.41 ±0.04 at 25°C

Catalog No. B82

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Each packet contains sufficient material to prepare 1 liter of solution with a pH of 7.41 ± 0.04 at 25°C
  • Convenient and ready-to-mix
  • Mix with deionized water to make 1L of pH buffer
  • Eliminates the need to store or carry bottles of buffer solution - simply transport the ready-to-mix packets
  • Intended for research use only; not for human or animal diagnostic purposes
Specifications

Specifications

Buffer Salt, pH 7.42
MFCD00050029
White
Certified
12 ea.
Envelopes
7558-79-4
Potassium Phosphate Monobasic/Sodium Phosphate Dibasic
Solid
7.41 ±0.04 (25°C)
Pass Test
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit