Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer pH 8.0, MilliporeSigma™
Supplier: MilliporeSigma BX16571
Specifications
Buffer pH 8.0 | |
Liquid | |
500 mL |
Safety and Handling
Recommended Storage : +15°C to +3°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction