Learn More
Chemglass Life Sciences Vapor Tube, Steam Duct, 160mm, 29/42 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 287mm Approx OAL
Supplier: Chemglass Life Sciences CG131731
Vapor Tube, Steam Duct, 160mm, 29/42 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 287mm Approx OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. Available in the 4 listed sizes. NOT SUPPLIED WITH COMBI-CLIP.Dimension “A”:CG-1317-31: 160mm