Learn More
Chemglass Life Sciences Bubbler, Horizontal, 24/40 Inner Joint, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF051402
Bubbler, Horizontal, 24/40 Inner Joint, Airfree, Schlenk. Horizontal design of bubbler allows for in line connection and purging of apparatus. Hose connection has a 9mm OD middle serration. Complete with standard taper inner joint.