Learn More
Chemglass Life Sciences Bubbler, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF051320
Bubbler, Airfree, Schlenk. Used to monitor gas flow to reactions while providing a vent to the atmosphere. Reservoir head prevents oil from being sucked back into the system. Volume below the sidearm is approx 40mL. Bubbler has an 8mm OD top and side tubulations.