Learn More
Chemglass Life Sciences Bubbler, Adjustable, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF0513A21
Bubbler, Adjustable, Airfree, Schlenk. Bubbler has an adjustable inner tube with a #9 Chem-Thread. Taper at bottom of reservoir allows for use of a minimal amount of oil. Complete bubbler consists of the lower reservoir, adjustable inner tube, compression cap and Viton o-ring.