Learn More
Sigma Aldrich Fine Chemicals Biosciences N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, 25561-30-2, MFCD00008269, 10x1 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1523810X1ML
Encompass
Linear Formula: CF3C[=NSi(CH3)3]OSi(CH3)3, Molecular Weight: 257.40, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS), Synonym: BSTFA + TMCS, Silylating mixture V.
Catalog No. NC1553105