Learn More
Brookfield Engineer Labs Brookfield Ametek Spindle RVHV #4 RVHV-4
Supplier: Brookfield Engineer Labs RVHV4
All standard RV, HA or HB torque Brookfield Ametek Viscometers/Rheometers are supplied with a standard spindle set (RV-2 through RV-7) constructed of #302 stainless steel. Additional spindle options are available in #316 stainless steel or Teflon coating for increased corrosion resistance.