Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Breastfeeding Chart



3B Scientific Breastfeeding Chart includes important information about breastfeeding, including its benefits.
Supplier: 3B Scientific™ 4006706
This item is not returnable.
View return policy
Description
- This colorful anatomical chart illustrates the basic anatomy of human breastfeeding.
- Important information about breastfeeding, including its benefits, are presented.
- The anatomy chart is complete with detailed instructions on the use of a breast pump.
- Printed on premium glossy (200 g) paper.
- Convenient Poster size 50 x 67 cm (20 x 26 in.).
Specifications
Breastfeeding Poster | |
Glossy Paper | |
Information About Basic Anatomy of Human Breastfeeding |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title