Learn More
Verichem Laboratories BR2 BILIRUBIN CALIBRATOR
Supplier:  Verichem Laboratories 9459
Encompass_Preferred
BR2 Bilirubin Calibrator. specifically designed for calibration of AAdvanced Instruments Model BR2 Bilirubin Stat-Analyzer. Protein based, ready-to-use liquid, no glycols, azides or surfactants, no special procedures or diluents required, stable 5 days after opening when refrigerated
Catalog No. NC0422992