Learn More
Eisco™ Boyle's Law Apparatus, Demonstration Type
SCI_ED
Wide-bore glass tube is closed at top and mounted on a scale.
Supplier:  Eisco™ PH0150
Catalog No. S99495
Add to Cart
Description
Description
Clearly illustrates Boyle's Law for students.
  • Wide-bore glass tube closed at top, mounted on a scale
  • Scale is 0 to 600mm with zero corresponding to the inside of the top end
  • Air is trapped by colored oil in a metal pressure chamber
  • Chamber fitted with a bourdon gauge and a riffled inlet tube with stopcock
  • Plastic safety screen over the tube
Specifications
Specifications
Clearly illustrates Boyle's law for students
Boyle's Law Demonstration Apparatus
2.82 kg
22 cm
8.66 in.
99.47 oz.
Metal pressure chamber fitted with bourdon gauge, inlet tube, stopcock, Plastic safety screen, wide bore tube mounted on a 0 to 600mm scale. (Supplied without oil and hand pump.)
15 cm
8.66 x 5.9 x 27.16 in. (22 x 15 x 69 cm)
69 cm
27.16 in.
5.9 in.
Product Suggestions
Product Suggestions