Learn More
JDM SCIENTIFIC LLC Box Scientific Heliport Plus Heated thawing station
Supplier: JDM SCIENTIFIC LLC 130071000
Heliport Plus has the same deck size as the Heliport but with the added feature of a 600 watt heating system that can be turned on to thaw large molecule compounds and high melt carriers such as DMSO. In addition, its combined with 7 upgraded fans that deliver 6 liters per minute of 30C air at 2 meters per second. The heater can be turned off when not needed for other compounds and will perform the same as the basic Heliport.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.